missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen Prepacked HIC Column, Capto Butyl

Intended for initial and intermediate step purifications that require a medium with low to medium hydrophobicity.

Brand:  Cytiva 28-9924-73

166.78 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



Product Code. 11320362

  • 180.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Capto Butyl often works efficiently with rather low salt concentrations
  • Small bed volume for fast results and minimal sample/buffer consumption
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Piece
Polypropylene
75μm (median particle size of the cumulative volume distribution)
HIC Column
<600cm/h
Butyl
4° to 30°C, 20% Ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen Capto Butyl, 1 x 4.7 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.