missing translation for 'onlineSavingsMsg'
Learn More
Histamine dihydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 10583311

Histamine dihydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 10583311
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10583311

Brand: Thermo Scientific Acros 150620050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5H11Cl2N3
Molecular Weight (g/mol) 184.06
MDL Number MFCD00012703
InChI Key PPZMYIBUHIPZOS-UHFFFAOYSA-N
Synonym histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PubChem CID 5818
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1

Specifications

Melting Point 249.0°C to 252.0°C
Color White
Loss on Drying 0.5% (105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 5 g
Beilstein 25315
Merck Index 154756
Solubility Information Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol
Formula Weight 184.07
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Histamine dihydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0400943 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150625000,150621000,150620250,150620050
Lot NumberA0398833 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number150625000,150621000,150620250,150620050
Lot NumberA0410033 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number150625000,150621000,150620250,150620050
Lot NumberA0412291 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number150625000,150621000,150620250,150620050
Lot NumberA0410875 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number150625000,150621000,150620250,150620050
compliance-icons compliance-icons
Product Identifier
  • Histamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Histamine dihydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.