Learn More
Epredia™ HistoScreen™ Cassettes
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
Brand:  Epredia™ B851000BE
 View more versions of this product
Product Code. 16323926
  • 677.00 € / Pack of 1000
Explore more special offers
Description
Description
  • Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
  • Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
  • Precision engineering for solid connection to microtomes.
  • Cassette surface texture optimizes scanability for archiving.
Specifications
Specifications
Mesh
1000/Cs.
Beige
Separate lids included
Back Hinge
HistoScreen™ Cassette