Learn More
Epredia™ HistoScreen™ Cassettes
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
Brand: Epredia™ B851000OR
495.00 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
- Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
- Precision engineering for solid connection to microtomes.
- Cassette surface texture optimizes scanability for archiving.
Specifications
Mesh | |
1000 | |
Orange |
Separate lids included | |
Back Hinge | |
Histoscreen™ Cassette |