missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HistoScreen™ Cassettes in Tube Packs

Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.

504.00 € - 528.00 €

Specifications

Closure Included Yes
Quantity 750/Cs.
Pore Style Mesh
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
View More Specs

Products 11
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16343936
View Documents
Epredia™
B851001BE
Beige
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16363936
View Documents
Epredia™
B851001GN
Green
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16373936
View Documents
Epredia™
B851001GY
Gray
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16383936
View Documents
Epredia™
B851001LA
Lavender
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16393936
View Documents
Epredia™
B851001LG
Light Green
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16303946
View Documents
Epredia™
B851001LI
Lilac
528.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16313946
View Documents
Epredia™
B851001OR
Orange
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16323946
View Documents
Epredia™
B851001PK
Pink
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16333946
View Documents
Epredia™
B851001TN
Tan
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16343946
View Documents
Epredia™
B851001WH
White
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
16353946
View Documents
Epredia™
B851001YW
Yellow
504.00 €
Pack of 750
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
Specifications

Specifications

Yes
Mesh
Yes
HistoScreen Cassette
750/Cs.
Separate Lid, Back Hinge
For Use With PrintMate™ Printers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ HistoScreen™ Cassettes in Tube Packs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.