Learn More
Simport™ Scientific Histosette™ I Biopsy Processing/Embedding Cassette
Designed to hold biopsy specimens during the embedding process. Histosette™ I Biopsy Processing/Embedding Cassette are suitable to be used with some models of cassette labeling instruments.
Specifications
Closure Included | Yes |
---|---|
Writing Face Angle | 45° |
Material | Acetal |
Product Type | Biopsy Cassette |
Description
- Made of high density polymer, acetal
- Keeps specimens safely submerged in liquid
- Resistant to chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely Available in 11 colors
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Yes | |
45° | |
Acetal | |
Biopsy Cassette |
For in vitro diagnostic (IVD) use only.