missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HisTrap™ FF Crude Prepacked Columns

Purify histidine-tagged proteins directly from homogenized, unclarified cell lysates. Cytiva Life Sciences™ HisTrap™ FF Crude Prepacked Columns offer the convenience of simple, one-step purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).

Brand:  Cytiva 17-5286-01

710.60 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



 View more versions of this product

Product Code. 11723219

  • 748.00 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Ni Sepharose™ 6 Fast Flow media
  • Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
  • 1 and 5mL columns

Media
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mL
16 mm
<20 mL/min.
3 to 12
4°C to 30°C
45 to 165 μm
Intended for research use only, and shall not be used in any clinical or in vitro procedures for diagnostic purposes
0.6 in.
HisTrap FF Crude columns are high binding capacity IMAC columns for his-tagged protein purification when scalability is needed.
Polypropylene
6% Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
5 x 5 mL
Pre-packed Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains Nickel. May produce an allergic reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.