missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HisTrap™ FF Crude Prepacked Columns
Purify histidine-tagged proteins directly from homogenized, unclarified cell lysates. Cytiva Life Sciences™ HisTrap™ FF Crude Prepacked Columns offer the convenience of simple, one-step purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
Brand: Cytiva 29048631
This item is not returnable.
View return policy
Description
- Prepacked with Ni Sepharose™ 6 Fast Flow media
- Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
- 1 and 5mL columns
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
1 mL | |
7 mm | |
<4 mL/min. | |
3 to 12 | |
4°C to 30°C | |
45 to 165 μm | |
Intended for research use only, and shall not be used in any clinical or in vitro procedures for diagnostic purposes |
0.2 in. | |
HisTrap FF Crude columns are high binding capacity IMAC columns for his-tagged protein purification when scalability is needed. | |
Polypropylene | |
6% Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
1 mL | |
Pre-packed Column |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.