missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Benzamidine FF (high sub) Columns
Remove or purify serine proteases such as thrombin. Cytiva Life Sciences™ HiTrap™ Benzamidine FF (high sub) Columns are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub).
219.00 € - 436.00 €
Specifications
Particle Size | 90 μm |
---|---|
For Use With (Equipment) | Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system |
Product Type | Benzamidine FF Column |
For Use With (Application) | Removal or purification of Serine Proteases such as Thrombin |
Height (English) | 0.98 in. |
Product Code | Brand | Quantity | Flow Rate | Diameter (Metric) Inner | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Flow Rate | Diameter (Metric) Inner | Price | Quantity & Availability | |||
11723199
|
Cytiva
17-5144-01 |
1 x 5mL | <20 mL/min. | 1.6cm |
428.00 €
Each |
In Stock
Log in to see stock. |
|||
10774357
|
Cytiva
17-5143-02 |
2 x 1mL | <4 mL/min. | 0.7cm |
219.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
10450135
|
Cytiva
17-5143-01 |
5 x 1mL | <4 mL/min. | 0.7cm |
436.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
Description
- Prepacked with Benzamidine Sepharose™ 4 Fast Flow (high sub) media
- Protein binding capacity ≥35mg trypsin/mL medium
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
Specifications
90 μm | |
Benzamidine FF Column | |
0.98 in. | |
>12μM p-aminobenzamidine/mL medium | |
42psi | |
4% Highly Cross-linked Agarose | |
2° to 8°C |
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system | |
Removal or purification of Serine Proteases such as Thrombin | |
2.5cm | |
Polypropylene | |
2 to 8 | |
Serine Proteases such as Thrombin |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Benzamidine FF (high sub) 5 ml, 1 x 5 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.