missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ Adhere Columns
Designed to remove contaminants in post-protein A purification
416.00 € - 981.00 €
Specifications
Buffer | Aqueous Buffer |
---|---|
Product Type | HiTrap Capto adhere column |
Height (English) | 0.98 in. |
Height (Metric) | 25mm |
Material | Polypropylene |
Product Code | Brand | Quantity | Diameter (English) Inner | Flow Rate | Max. Pressure | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Diameter (English) Inner | Flow Rate | Max. Pressure | Price | Quantity & Availability | ||
10635935
|
Cytiva
28-4058-44 |
5 x 1mL | 0.30 in. | <4 mL/min. | 1.5 bar |
416.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||
11505135
|
Cytiva
28-4058-46 |
5 x 5mL | 0.62 in. | <20 mL/min. | 44 bar |
981.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||
Description
- Prepacked with Capto adhere media, a strong anion exchanger with multimodal functionality
- High capacity and productivity in flow-through mode: 0.09 to 0.12mmol Cl-/mL medium ionic capacity
- Wide operational window of pH and conductivity: working pH range 3 to 12
- Two-step process reduces time spent and operating costs
Specifications
Aqueous Buffer | |
0.98 in. | |
Polypropylene | |
3 to 12 | |
Highly Cross-linked Agarose | |
Affinity Column |
HiTrap Capto adhere column | |
25mm | |
N-benzyl-n-methyl Ethanolamine | |
75μm | |
4° to 30°C | |
Easy screening and convenient process development |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Capto adhere 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title