missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Core 400

Designed for intermediate purification and polishing of viruses and other large biomolecules

Brand:  Cytiva 17372411


Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15948862

  • 467.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • An excellent alternative to size‐exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacturing
  • Novel core bead technology and multimodal ligands provide dual functionality, size exclusion, and binding chromatography
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
  • High capacity and productivity in flowthrough mode
  • Wide operational window of pH and conductivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5/Pk.
Intermediate Purification, and Polishing of Viruses and Large Biomolecules
Capto™ Core 400
Prepacked HiTrap™ Column
1 mL
0.5 MPa (5 bar)
HiTrap™
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P235-Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers