missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ IEX Selection Kit
Designed for use with syringe, peristaltic pump, or chromatography systems such as AKTA design for convenient media screening and optimization studies
Brand: Cytiva 28-9343-88
This item is not returnable.
View return policy
Description
- Consists of five HiTrap columns, prepacked with Capto ion exchange chromatography media with different characteristics
- For media screening; includes five 1mL HiTrap columns prepacked with Capto Q, Capto S, Capto DEAE, Capto MMC, Capto adhere, as well as connectors and instructions
- For optimization studies; buffer composition, pH, flow rate, sample loading and elution schemes can be optimized with small sample quantities prior to scale-up
- Capto Q is strong anion exchanger, Capto DEAE is weak anion exchanger, Capto S is strong cation exchanger
- Capto MMC and Capto adhere media have multimodal characteristics that can be used to solve specific purification problems
- Capto MMC is multimodal cation exchanger that is functional at high as well as at low ionic strength
- Capto adhere is multimodal strong anion exchanger mainly designed for post-protein A purification of monoclonal antibodies
- Prepacked columns and bulk media are available for larger scale preparative purifications
- Max. pressure (over packed bed during operation): 3 bar (42psi)
- Flow rate: <4mL/min.
- Complete packsize: 5 x 1mL
- Bed dimensions: 7 × 25mm
- Bed volume: 1mL
- pH Stability cleaning: 2 to 14
- Matrix: highly cross-linked agarose with dextran surface extender
- Average particle size: 90μm (d50V)
- pH Stability working range: 2 to 12
- Pressure/Flow specification: 0.3MPa at 700cm/hr., 1m diameter column, 20cm bed height
Column
- Column I.D.: 7mm
- Material (column hardware): polypropylene (PP)
Specifications
Ion Exchange Chromatography | |
HiTrap Capto IEX Selection Kit | |
7 mm | |
Syringe, peristaltic pump and other chromatography systems such as AKTAdesign |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Capto IEX Selection Kit
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction