missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto MMC Columns

HiTrap Capto MMC is prepacked with Capto MMC, a multimodal BioProcess ion exchange medium. The multimodal functionality gives a different selectivity compared with traditional ion exchangers and it binds proteins at high or low ionic strength.

289.00 € - 686.00 €

Specifications

Buffer Aqueous Buffer
Product Type HiTrap Capto MMC column
Height (English) 0.98 in.
Height (Metric) 25mm
Material Polypropylene
View More Specs

Products 2
Product Code Brand Quantity Diameter (English) Inner Flow Rate Max. Pressure Price Quantity & Availability  
Product Code Brand Quantity Diameter (English) Inner Flow Rate Max. Pressure Price Quantity & Availability  
10480315
View Documents
Cytiva
11-0032-73
5 x 1mL 0.27 in. <4 mL/min. 44 bar
289.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
11748408
View Documents
Cytiva
11-0032-75
5 x 5mL 0.62 in. 300kPa at 600cm/hr. 3 bar
686.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

  • High dynamic binding capacity at high conductivity.
  • Selectivity allows binding in the presence of salt.
  • High volume throughput.
  • HiTrap Capto MMC can be used to bind proteins at the conductivity of the feed material and to solve specific purification challenges.
Specifications

Specifications

Aqueous Buffer
0.98 in.
Polypropylene
75μm
4° to 30°C
Bind Proteins at the conductivity of the feed material and to solve specific purification challenges
HiTrap Capto MMC column
25mm
2 to 12
Highly Cross-linked Agarose
Affinity Column
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hitrap capto MMC

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.