missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ Q IEX Columns
Strong quaternary ammonium (Q) anion exchanger
Brand: Cytiva 11-0013-03
This item is not returnable.
View return policy
Description
- High-throughput medium for capture and intermediate purification
- For process-scale purification
- Combined properties of high binding capacity, high flowrate and low back-pressure properties reduce process cycle times and improve productivity
- Rigid agarose matrix enables high bed heights and purification of viscous samples at high flowrates
- High dynamic binding capacity is achieved through dextran surface extender coating on the agarose matrix
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
HiTrap Capto Q | |
0.6 in. | |
<20 mL/min. | |
Quaternary amine | |
16 mm | |
90 μm (d50V) | |
5 x 5 mL |
Polypropylene | |
Highly cross-linked agarose with dextran surface extender | |
HiTrap Capto Q chromatography column is prepacked with a strong anion exchange modern resin for small scale protein purification. | |
4°C to 30°C in 20% Ethanol | |
5 mL | |
2 to 12 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- hitrap capto Q 5ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction