missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns

Product Code. p-4907650 Shop All Cytiva Products
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11673074

Brand: Cytiva 17547055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-throughput medium for intermediate and polishing protein purification

  • Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
  • Screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Strong quartenary amine anion exchanger

TRUSTED_SUSTAINABILITY

Specifications

Description HiTrap Capto Q ImpRes, 5 x 5 mL
Flow Rate <20 mL/min.
For Use With (Application) HiTrap Capto Q ImpRes chromatography column is packed with a strong anion exchange modern resin for high resolution, small-scale protein purification.
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 7 mm
Volume (Metric) 5 mL
Quantity 5 x 5 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.