missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Con A 4B Columns
For separation and purification of glycoproteins, polysaccharides and glycolipids
216.00 € - 803.00 €
Specifications
Buffer | Aqueous Buffer |
---|---|
Product Type | HiTrap Con A Sepharose 4B column |
Height (English) | 0.98 in. |
Height (Metric) | 25mm |
Material | Polypropylene |
Product Code | Brand | Quantity | Diameter (English) Inner | Flow Rate | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Diameter (English) Inner | Flow Rate | Price | Quantity & Availability | |||
11721417
|
Cytiva
28-9520-85 |
5 x 1mL | 0.27 in. | <4 mL/min. |
216.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
11731417
|
Cytiva
28-9520-96 |
5 x 5mL | 0.62 in. | <20 mL/min. |
803.00 €
Pack of 5 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
Description
- Prepacked with Con A Sepharose™ 4B media
- Bed volume: 1mL and 5mL
- Bed dimensions: 7 x 25mm (1mL), 16 x 25mm (5mL)
- Column material: Polypropylene (PP)
- Max flow rate: 4mL/min (1mL), 20mL/min (5mL)
- Ligand: Concanavalin A (Con A)
- Matrix: 4% agarose
- Average particle size: 90μm
- pH stability working range: 4 to 9
Specifications
Aqueous Buffer | |
0.98 in. | |
Polypropylene | |
3 bar | |
45 to 165μm | |
4° to 8°C | |
Convenient separation and purification of Glycoproteins, Polysaccharides and Glycolipids |
HiTrap Con A Sepharose 4B column | |
25mm | |
Concanavalin A (13mg Con A/mL drained medium) | |
4 to 9 | |
4% Agarose | |
Ion Exchange Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Con A 4B 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title