missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ DEAE Sepharose Fast Flow IEX Columns

Weak anion exchanger for small-scale protein purifications. Cytiva Lifescience™ HiTrap™ DEAE Sepharose Fast Flow IEX Columns are convenient and rapid.

Brand:  Cytiva 17-5154-01

344.85 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 11515025

  • 374.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HiTrap DEAE FF
<20 mL/min.
4°C to 30°C in 20% Ethanol
5 mL
0.6 in.
For method scouting, group separations, sample concentration and sample clean-up of charged biomolecules
16 mm
5 x 5 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap DEAE FF 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.