missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ IEX Selection Kit

Use for fast and easy screening of a variety of different media. HiTrap™ IEX Selection Kits contain Sepharose™ Fast Flow and Sepharose XL in combination with seven different ion exchange ligands.

Brand:  Cytiva 17-6002-33


Product Code. 10186074

  • 315.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Seven prepacked 1mL HiTrap columns, with one each of the following: Q Sepharose Fast Flow (strong anion), SP Sepharose Fast Flow (strong cation), CM Sepharose Fast Flow (weak cation), DEAE Sepharose Fast Flow (weak anion), ANX Sepharose 4 Fast Flow (high sub) (weak anion), Q Sepharose XL (strong anion), and SP Sepharose XL (strong cation); connectors; and instructions
This item is not returnable. View return policy

Description

Description

  • Broad array of weak/strong anion and cation exchangers prepacked in 1mL HiTrap columns allow screening of a variety of different media
  • Excellent for optimization studies: buffer composition, pH, flowrate, sample loading, and elution scheme can be optimized with small sample quantities prior to scale-up
  • Designed for use with syringe, peristaltic pump, or chromatography system
  • Recommended column flowrate, <4mL/min.; maximum pressure: 3 bar

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HiTrap IEX Selection Kit
<4 mL/min.
4°C to 30°C, 20% Ethanol (Q, DEAE, ANX, cm), 20% Ethanol, 0.2 M Sodium Acetate (SP)
1 mL
0.2 in.
HiTrap IEX Selection Kit is used for screening of the optimal GE ion exchange resin for specific application and development work.
7 mm
7 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap IEX Selection Kit'

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, ethanol, ethanol, ethanol, ethanol, ethanol, ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.