missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ IgG Sepharose™ FF

For use in chromatography. Cytiva Lifescience HiTrap™ IgG Sepharose™ FF is stable to all commonly used aqueous buffers.

Brand:  Cytiva 90100397


Product Code. 15599271

  • 1449.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • pH Stability Working Range 3–10
  • Pressure/Flow Specification: Base matrix: 250-400cm/h, 100kPa, XK 50/60 column, bed height 25cm
  • Bed Dimensions: 7mm × N/A
  • Column i.d.: 7mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

90 μm
pH Stability Working Range 3-10, pH Stability Cleaning 3-10, pH stability, CIP 3-10
Packing Material
Polypropylene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers