missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap MabSelect PrismA Column
Protein A Column
227.00 € - 4269.00 €
Specifications
Max. Operating Pressure | 5 bar (0.5 MPa, 70 psi) |
---|---|
For Use With (Application) | Antibody affinity chromatography |
Type | Pre-packed Column |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15803975
|
Cytiva
17549851 |
1 x 1 mL |
227.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15813975
|
Cytiva
17549852 |
5 x 1 mL |
895.00 €
Pack of 5 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
15823975
|
Cytiva
17549853 |
1 x 5 mL |
933.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15833975
|
Cytiva
17549854 |
5 x 5 mL |
4269.00 €
Pack of 5 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
5 bar (0.5 MPa, 70 psi) | |
Antibody affinity chromatography | |
Pre-packed Column |