missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Macrocap™ SP

For use in method scouting, group separations, sample concentration and sample clean-up of charged biomolecules. Cytiva Lifescience HiTrap™ Macrocap™ SP are made of biocompatible polypropylene that does not interact with biomolecules.

Brand:  Cytiva 28950859

Product Code. 15569244

  • 2080.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • BioProcess resin
  • pH Stability Cleaning 2–13
  • pH stability, CIP 2–13
  • Chemical Stability All commonly used aqueous buffers, 0.5 M NaOH, 0.1 M citric acid, 25% ethanol, 30% propanol, 30% methanol, 50% ethylene glycol, 1% Tween-20, and 1% SDS
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
Acrylamide-dextran copolymer
SO3−
16 mm
50 μm
0.6 in.
Method scouting, group separations, sample concentration and sample clean-up of charged biomolecules
4°C to 30°C, 20% Ethanol, 0.2 M Sodium Acetate
5 mL
pH Stability Working Range: 4 to 11
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HiTrap™ Macrocap™ SP > HiTrap Macrocap SP

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.