missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Protein G HP Prepacked Columns
Use for fast, easy separations of IgG from different species and subclasses. Cytiva Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.
Brand: Cytiva 29048581
228.95 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Prepacked with Protein G Sepharose High Performance
- Binding capacity: 25mg human IgG/column
- Maximum pressure: 3 bar
- Column material: polypropylene
- For use with syringe, peristaltic pump, or chromatography system
- 1 and 5mL columns
Media
- Ligand: Recombinant protein G lacking albumin-binding region
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: 3 to 9
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Pre-packed Columns | |
7 mm | |
<4 mL/min. | |
Designed for use with a syringe, peristaltic pump, or chromatography system | |
20% Ethanol | |
1 Pack |
25 mg Human IgG/Column | |
0.27 in. | |
1 mL | |
HiTrap Protein G HP are prepacked columns for general-purpose preparative purification of monoclonal and polyclonal antibodies from most species including rat. | |
4°C to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur