missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Q Sepharose XL IEX Columns
Optimized for fast, convenient small-scale protein capture
Brand: Cytiva 17-5158-01
158.65 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed for use wtih a syringe, peristaltic pump or chromatography system
- For group separations, concentrations and sample cleanup
- Convenient and affordable for fast, easy separations either alone or connected in series
- High binding capacities with high flowrates
- Media consists of long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix
- Dextran chains increase the exposure of charged groups for high loading capacities
Specifications
HiTrap Q XL | |
<4 mL/min. | |
4°C to 30°C in 20% Ethanol | |
1 mL |
0.2 in. | |
HiTrap Q XL are strong anion exchangers prepacked with Q Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography. | |
7 mm | |
5 x 1 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiTrap Q XL 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol