missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HM 430, HM 450 and 440 E Sliding Microtome Accessories

Maintain Epredia™ HM 430 and HM 450 sliding microtomes with Epredia™ HM 430, HM 450 and 440 E sliding microtomes accessories.

POA - 1465.00 €

Specifications

Product Type Sliding Microtome Accessories
For Use With (Equipment) Epredia HM 430 and HM 450 Sliding Microtomes
View More Specs

Products 8
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12093019
Encompass
View Documents
Epredia™
152160
Wooden Case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12043049
View Documents
Epredia™
440400
Height Adjustment Plate, 10 mm
83.05 €
Each
Please to purchase this item. Need a web account? Register with us today!
12053049
View Documents
Epredia™
440410
Height Adjustment Plate, 20 mm
95.35 €
Each
Please to purchase this item. Need a web account? Register with us today!
12003059
View Documents
Epredia™
502140
Waste Tray This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12073099
missing translation for 'viewDocuments'
Epredia™
705200
Blade Holder (for insertion of low-profile blades)
1465.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12093099
Encompass
missing translation for 'viewDocuments'
Epredia™
705430
Disposable Blade Holder This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12003109
Encompass
missing translation for 'viewDocuments'
Epredia™
705440
Low-profile Disposable Blade Holder, 2 blades This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12033109
Encompass
missing translation for 'viewDocuments'
Epredia™
705490
Blade Holder Case This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschrijving

Beschrijving

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
  • Cooling element for buffer solution

  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
  • Large-field magnifier

  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
  • StemiScope™ with optics and mount

  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
  • Swan-neck light guide

  • 3.5 mm fiber bundle
  • Swan-neck light guide: 50 mm long
  • Flexible light guide: 750 mm long
  • High light intensity
  • Include adapter for the KL200 (760317)
  • Specificatie

    Specificatie

    Sliding Microtome Accessories
    Epredia HM 430 and HM 450 Sliding Microtomes
    Video's
    SDS
    Documentatie

    Documentatie

    Certificates