missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HM 650V Vibrating Blade Microtome Accessories

Product Code. 12093039 Shop All Epredia Products
Click to view available options
Product Type:
Cyanacrylat Glue
Microtome Blade Holder
Unit Size:
20mL
Each
This item is not returnable. View return policy

Product Code. 12093039

Brand: Epredia™ 350220

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Standard buffer tray

  • Holds stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier
  • Magnification 2.5X
  • Move, swivel, and displace for optimal viewing position
  • Includes adapter and illumination
StemiScope™ with optics and mount
  • Eyepieces: 10X
  • Supplementary lens: 0.63X
  • Zoom magnifying range: 4.1 to 31.5X
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide
  • 3.5mm fiber bundle
  • Swan-neck light guide: 50mm long
  • Flexible light guide: 750mm long
  • High light intensity
  • Include adapter for the KL200 (760317)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cyanacrylat Glue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.