Learn More
Novus Biologicals™ HMGCL Overexpression Lysate
Brand:  Novus Biologicals™ NBL1-11617
Product Code. 18225855
  • 700.00 € / 0.10mg
Explore more special offers
Description
Description
The HMGCL Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC23817 and based on accession number NM_191. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
Western Blot
RIPA buffer
Homo sapiens 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase (HMGCL), mRNA.
Cancer, Signal Transduction