missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16898-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
HMGCL | |
Polyclonal | |
Unconjugated | |
HMGCL | |
HL, HMG CoA lyase, HMGCL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15356, 3155, 79238 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35914, P38060, P97519 | |
HMGCL | |
HMGCL Fusion Protein Ag10426 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |