Learn More
Invitrogen™ Hoechst 33342 Ready Flow™ Reagent
Hoechst 33342 Ready Flow™ Reagent is a bright, easy-to-use cell-permeant stain for cell cycle analysis on a flow cytometer.
Marke: Invitrogen™ R37165
240.35 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Beschreibung
This dye emits blue fluorescence when bound to double-stranded DNA with an emission maximum at 461 nm.
• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Cell-permeable and effective in fixed and live-cell staining for cell cycle
Hoechst 33342 Ready Flow Reagent is a cell-permeable, blue fluorescent DNA stain for cell cycle analysis. Add 2 drops from the dropper bottle directly into 1 x 106 cells, incubate, and analyze.
Hoechst 33342 reagent binds preferentially to adenine-thymine regions of double-stranded DNA. This dye binds into the minor grooves of DNA and exhibits distinct fluorescence emission spectra that are dependent on dye:base pair ratios. Hoechst 33342 Ready Flow Reagent is excited by UV light and has peak emission at 460 nm.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
Blue | |
361/497 | |
120 assays | |
Nucleic Acid Stain |
2 dropper bottles, store at room temperature | |
Cell-Permeant | |
Ready Flow™ | |
Nucleus |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.