Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand:  Hoefer™ TE26
 View more versions of this product
Product Code. 11544434
  • 76.45 € / Pack of 50
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Western Blotting
Specifications
Specifications
9 cm
TE22 Mini Tank Blotting Unit or MiniVE Blot Module
Cellulose
10.5 cm
Sheet