missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Brand:  Hoefer™ SE255

 View more versions of this product

Product Code. 12024877

  • 123.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Designed for quick screening and rapid separation
Specifications

Specifications

SE 250
Buffer Chamber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers