Learn More
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Film
For use with the SE 1200 Easy Breeze Gel Drying System
Brand: Hoefer™ SE1202
Code : 04
Additional Details : Weight : 1.00000kg
Description
Gel Drying Film
- For photography or fluorography, clear cellophane ensures a photograph or exposure without distortion
- For autoradiography, use of a Mylar™ sheet in place of one cellophane sheet allows the Mylar to be easily removed after drying for maximum beta emission
Specifications
Standard Frame of the SE 1200 Easy Breeze™ Air Gel Drying System | |
Porous Cellophane Sheet |