Learn More
Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Filter Paper Sheet
Dries acrylamide and agarose gels rapidly, evenly and safely.
Brand: Hoefer™ SE1141
Code : NEW
Additional Details : Weight : 1.22000kg
Description
- 35 x 44 cm
- 25 sheets
Specifications
GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers | |
Filter Paper sheet |