missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ HE 33 Mini Submarine Electrophoresis Unit
Designed for very fast separations of DNA restriction fragments in agarose gels
Brand: Hoefer™ HE33-8-1.5
Code : 04
Weitere Details : Weight : 0.76500kg
Beinhaltet: Buffer chamber, lid with cables, UV-transparent gel running tray, gel casting tray, 4 foam gaskets, bubble level. Complete system (03-500-130) also includes 1.5mm thick 8-well comb and comb back.
Beschreibung
Designed for very fast separations of DNA restriction fragments in agarose gels
- The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
- Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
- No taping required for casting
- Electrical safety locks in lid
Spezifikation
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
500 mA | |
Up to 80% | |
Up to 32 | |
50°C | |
250 mL | |
9.4 in. | |
24 cm | |
5.1 in. | |
13 cm | |
2.8 in. | |
500 V | |
500 mA |
Mini | |
Mini Horizontal Agarose Electrophoresis Unit | |
Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level, 1.5mm thick, 8-well comb, comb back | |
122°F | |
8.4 oz. | |
9.4 x 5.1 x 2.8 in. (24 x 13 x 7 cm) | |
3.9 in. | |
10 cm | |
2.7 in. | |
7 cm | |
7 cm | |
15 w |