Learn More
Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
Brand:  Hoefer™ HE99X
 View more versions of this product
Product Code. 11519573
  • 553.00 € / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Includes:
Buffer chamber with electrodes and leveling feet, safety lid with leads, bubble level. Complete system (03-500-121) also includes 15 x 20cm gel casting tray and UV-transparent running tray; 1.5mm thick, 15-well comb; adjustable comb back that permits control of the depth of the sample wells
Description
Description
  • Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
  • Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
  • Rugged design provides reliability and safety
Specifications
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Up to 80%
60
45°C
1200 mL
14.1 in.
36 cm
7.1 in.
18.2 cm
5.5 in.
200 V
100 mA
Horizontal Agarose Electrophoresis Unit
Buffer chamber assembly, safety lid with high voltage Leads, Foam gaskets, bubble level
113°F
40.5 oz.
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm)
5.9, 3.9 or 7.8 in.
15, 10 or 20 cm
5.9 in.
15 cm
14 cm
20 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products