missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Mini and Standard Electrophoresis Blotting Systems

Product Code. 11955388
Click to view available options
Includes:
Mini Blot Unit with Semidry Unit
Mini Blot Unit with Transfer Tank
Std Blot Unit with Semidry Unit
Std Blot Unit with Transfer Tank
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11955388

Brand: Hoefer™ 360622

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

The Mini Electrophoresis Blotting System with Transfer Tank includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; TE22 Mini Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Mini Electrophoresis Blotting System with Semidry Unit includes the Mighty Small II Mini Vertical Electrophoresis System, Model SE260 Complete; Semidry Transfer Unit with Built-In Power Supply, Model TE70XP; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Transfer Tank includes the SE600 Chroma Vertical Electrophoresis System; TE62 Tank Blotting Unit; and PS300B Power Supply (300V, 500mA, 90w). The Standard Electrophoresis Blotting System with Semidry Unit includes the SE600 Chroma Vertical Electrophoresis System; Large Semidry Transfer Unit with Built-In Power Supply, Model TE77XP; and PS300B Power Supply (300V, 500mA, 90w).

Economical equipment packages provide proven electrophoresis solutions for Western blotting

  • Comes with PS300B 300 V, 500 mA, 90 W Power Supply for all units
  • Save time and money with a combination package

Specifications

Content And Storage Deluxe dual cooled vertical unit, standard transfer tank with cooling chamber, power supply
Type Standard Electrophoresis Blotting System
Includes Std Blot Unit with Transfer Tank
Voltage 300 V
Wattage 90 w
Amperage 500 mA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.