Learn More
Hoefer™ MiniVE Mini Vertical Electrophoresis Unit
Accepts gels up to 10 x 10.5cm, increasing separation length by more than 20% over conventional 10 x 8cm gels
Brand: Hoefer™
Code : NEW
Additional Details : Weight : 1.00000kg
Includes:
Tank with lid, three 10 x 10.5cm rectangular glass plates, three 10 x 10.5cm notched glass plates, two gel modules, two 1mm thick 10-well combs, two 1mm thick spacer sets
Description
- Gel module is simultaneously a casting stand and an upper buffer chamber
- Cooling system assures the temperature uniformity required for straight lanes and flat bands
- Optional blot module (Mfr. No. SE302) permits gels to be cast, run and blot in one instrument
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
Lower Buffer chamber (tank), gel modules with gaskets-2 assemblies, safety lid with high voltage Leads, rectangular Glass plates, notched Glass plates, combs, T-shape spacers, well locating decals, wonder wedge plate separation tool, gel seal | |
45°C | |
0.03 in. | |
47.33 oz. | |
7.55 in. | |
19.2 cm | |
6.77 in. | |
17.2 cm | |
18.8 cm | |
25 w |
Up to 80% | |
113°F | |
15°C to 30°C | |
0.1 cm | |
1400 mL | |
3.93 in. | |
10 cm | |
3.14/4.1 in. | |
7.4 in. | |
500 V |