Learn More
Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates
Product Code. 11549693
Click to view available options
Type:
Low Fluorescence Notched Plate
Low Fluorescence Plate
Notched Plate
Material:
Alumina
Glass
Unit Size:
Pack of 5
Product Code. 11549693
Brand: Hoefer™ SE262P5
Only null left
Add to basket
Add to basket
Use with SE 260 or miniVE vertical electrophoresis units. Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
  • Size: 10 x 10.5 cm
Specifications
For Use With (Equipment) SE 260, MiniVE™ Vertical Electrophoresis Unit
Type Low Fluorescence Plate
Material Glass
Shape Rectangular