Learn More
Hoefer™ SE 600 Series Complete Vertical Electrophoresis System
Built-in heat exchanger maintains temperature between 1° and 45°C (33.8° and 113°F) for faster runs with superior quality results
Brand:  Hoefer™ SE600-15-1.5
Product Code. 10321575
  • 1955.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Offers durable construction for proven decades of performance
  • Run one gel or two gels simultaneously, four with optional divider plates
  • Produce straight lanes and sharp, well-defined bands—no “smiling”
  • Provide two mechanisms to dissipate heat
  • Gel sandwiches are completely immersed in the lower buffer so that heat is transferred evenly on both sides of the gel
  • A heat exchanger suspended vertically in the lower chamber cools or heats buffer evenly
Suitable for denaturing and native polyacrylamide, 2-D, isoelectric focusing and agarose gel electrophoresis
Certifications
UL3101-1, CSA C22.2 1010.1, CE Certified, EN61010-1
Specifications
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Lower Buffer chamber, upper Buffer chamber, safety lid with high voltage Leads, heat exchanger, Glass plates, clamp assemblies, cams, slotted gaskets for upper Buffer chamber, dual gel casting stand with leveling base, bubble level, laminated gaskets for
45°C
0.05 in.
169.07 oz.
12.59 in.
32 cm
11.41 in.
29 cm
50 w
Up to 80%
113°F
15°C to 30°C
0.15 cm
5000 mL
7.08 in.
18 cm
6.29/3.14 in.
1000 V
500 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products