Learn More
Hoefer™ SE 900 Large Format Vertical Electrophoresis System
Ideal for use with 24cm IPG strips in the second dimension of 2-D electrophoresis
Brand:  Hoefer™ SE900-1.0
 View more versions of this product
Product Code. 11995368
  • 8630.38 € / Each
Explore more special offers
Includes:
  • Buffer chamber with pumps, safety lid with leads, gel rack, 10 ft. tubing, two pair recirculation bath connectors, drain connector, seven tubing clamps, gel seal, wonder wedge plate separation tool
  • Mfr. No. SE900-1.0 also includes: Multicaster with ported face plate, six red spring clamps, two thumb screws, ten divider sheets, filler plug, cassette blank, gel seal, six glass hinge casettes
Description
Description
  • Runs 1 to 6 gels
  • Uses hinged cassettes for convenience and straight dye fronts
  • Easy to use — no clamps, no loose parts, bottom drain port
  • Utilizes internal pump for even temperature distribution
  • Compatible with a recirculating chiller for active cooling
Certifications
UL61010-1, CSAC22.2 No. 61010-1, EN61010-1:2001, CE Marked
Specifications
Specifications
EN61010-1, UL61010-1, CSA C22.2, CE
Separation tank, tubing kit, lid with high voltage Leads, multiple gel caster, internal PAGE rack, hinged Glass cassettes
45°C
16.92 in.
43 cm
16.92 in.
43 cm
20 cm
13.6 kg
150 w
Up to 80%
113°F
15°C to 30°C
9.84 in.
25 cm
7.87 in.
7.87 in.
29.98 lb.
600 V
1000 mA