Learn More
Hoefer™ TE22 Mini Tank Blotting Unit
Safe, economical unit requires minimal current and only about 1L of buffer
Brand:  Hoefer™ TE22
Product Code. 10380895
Quantity Price
1 1265.00 € / Each
3 1071.46 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Buffer chamber, lid with leads, two electrode panels, four gel cassettes, eight 0.13 in. thick foam sponges, four 0.25 in. thick foam sponges, 25 sheets blotting paper
Description
Description
  • Built-in alumina heat exchanger permits efficient transfers in less than one hour
  • Platinum electrode ensures uniform electrical field for complete and even transfers
  • Hinged, color-coded cassettes enable simple loading and easy orientation of gel sandwich
  • Alumina-covered serpentine channel in base provides excellent temperature control with no more than 5°C rise during a normal run
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Mighty Small™ Transfer Tank
Lower Buffer chamber with Alumina heat exchanger, safety lid with high voltage Leads, two electrode panels, four gel cassettes, eight 0.13 in. thick Foam sponges, four 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook
45°C
≈1000 mL
6.49 in.
5.51 in.
9.44 in.
100 V
500 mA
Up to 4, 9 x 10 cm Gel
Up to 80%
113°F
≈33.81 oz.
6.49 x 5.51 x 9.44 in. (16.5 x 14 x 24 cm)
16.5 cm
14 cm
24 cm
200 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products