Learn More
Hoefer™ Vacuum Gel Dryer
The user can set independent timers for heat and for vacuum
Brand: Hoefer GD2002
Code : 04
Additional Details : Weight : 5.00000kg
Description
The user can set independent timers for heat and for vacuum. The thermostat is adjustable from 40°C to 80°C, ±2°C - temperatures that vaporise water but do not burn the gels- Dries acrylamide and agarose gels up to 330mm x 440mm in as little as 30min
- The heavy aluminum platen provides even heat distribution and even vacuum distribution under the gel
- A clear silicone cover sheet allows the user to view the gel during drying
- The quiet, oil-free diaphragm pump maintains the low vacuum necessary for rapid gel drying. Requires no oil and no trap
Specifications
Vacuum Gel Dryer | |
230 V | |
50/60 Hz |