missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ HOFFMANN™ Pinch Cock Tube Clamps

Pinch cock tube clamps

Brand:  Bochem™ 12530

 View more versions of this product

Product Code. 11875091

  • 46.65 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pinch cock type
  • Includes flap
Specifications

Specifications

10/Pk.
Ms-Ni
12 mm
Pinchcock Tube Clamp
For pipes up to O.D. 40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers