missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Holder Clamp

OHAUS™ Holder Clamp provides secure support for holding glassware at elevated positions.

Brand:  OHAUS™ 30400045

 View more versions of this product

Product Code. 15772459

  • 103.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel is autoclavable and corrosion-resistant
  • Aluminum is lightweight yet strong
  • Cast iron with black enamel finish is chemical-resistant
  • Available in different shapes and sizes to fit your laboratory needs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fixed 90° Connection Angle
Aluminum
102.000 mm
6 to 24 mm
102.00 mm
Holder
Clamp, Holder, Ultra Clamp, CLC-ULTRAA
4.00 in.
4.00 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers