Learn More
SCHOTT AG Lighting and Imaging™ Holder Clamp for Fiber Optic Light Guides or LED Ring Lights
Holder Clamp to position fiber optic light guides or LED Ring Lights precisely. For use with an articulating arm (158345) and base (158 340).
Brand:  SCHOTT AG Lighting and Imaging™ 158341
 View more versions of this product
Product Code. 15713032
  • 79.00 € / 1 unit
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holder clamp to position fiber optic flexible light guides or Ring Lights precisely.
  • 1 x M6 hole.
  • Articulating arm and base available separately.
  • Holders for different size light guides and focusing lenses.
Specifications
Specifications
Holder Clamp
5 mm
Focusing Lens for Flexible Lightguides up to 5 mm dia.
Articulating arm 158 345 and optional a base 158 340
0.2 in.
Stereo Microscopy Illumination
1 M6 hole