missing translation for 'onlineSavingsMsg'
Learn More

Holmium foil, 0.127mm (0.005in) thick, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11385689
missing translation for 'orderingAttributeHoverText'
Quantity:
25 x 25 mm
50 x 50 mm
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Form: Approx.0.70g/25 x 25mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Especificaciones

Color Silver to Gray
Physical Form Foil
Quantity 50 x 50 mm
Assay Percent Range 99.9% (REO)
Odor Odorless
Appearance Silver gray foil
Chemical Name or Material Holmium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'