missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11383457
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Holmium(III) chloride has many application in the field of nuclear industry, lasers research applications. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-62-2
Molecular Formula Cl3Ho
Molecular Weight (g/mol) 271.28
MDL Number MFCD00011051
InChI Key PYOOBRULIYNHJR-UHFFFAOYSA-K
Synonym holmium chloride, holmium trichloride, holmium chloride hocl3, holmium iii chloride, wln: ho n-o3*3, holmium iii chloride, anhydrous, holmium iii chloride, anhydrous, ?hocl3, holmium iii chloride, anhydrous, powder trace metals basis, 3ar,8ar---4,4,8,8-tetrakis 3,5-diethylphenyl tetrahydro-2,2-dimethyl-6-phenyl-1,3-dioxolo 4,5-e dioxaphosphepin
PubChem CID 24992
SMILES [Cl-].[Cl-].[Cl-].[Ho+3]

Specifications

Melting Point 718°C
Quantity 5 g
Merck Index 14,4727
Solubility Information Soluble in water.
IUPAC Name holmium(3+) trichloride
Formula Weight 271.29
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.7 g/mL
Assay (metals basis)
Chemical Name or Material Holmium(III) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'