missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

158.79 € - 448.23 €

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
MDL Number MFCD00011053
InChI Key OWCYYNSBGXMRQN-UHFFFAOYSA-N
IUPAC Name diholmium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ho+3].[Ho+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311009
View Documents
Thermo Scientific Alfa Aesar
011281.09
10 g
158.79 €
10g
Please to purchase this item. Need a web account? Register with us today!
11301009
View Documents
Thermo Scientific Alfa Aesar
011281.18
50 g
448.23 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12055-62-8
377.86
OWCYYNSBGXMRQN-UHFFFAOYSA-N
[O--].[O--].[O--].[Ho+3].[Ho+3]
Ho2O3
MFCD00011053
diholmium(3+) trioxidandiide

Specifications

2360°C
Beige
99.99% (REO)
MFCD00011053
OWCYYNSBGXMRQN-UHFFFAOYSA-N
diholmium(3+) trioxidandiide
377.86
Holmium(III) oxide
12055-62-8
Powder
Ho2O3
Insoluble in water. Soluble in perchloric acids.
[O--].[O--].[O--].[Ho+3].[Ho+3]
377.86
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-015-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.