Learn More
Holmium(III) oxide, REacton™, 99.995% (REO), Thermo Scientific Chemicals
59.50 € - 664.00 €
Chemical Identifiers
CAS | 12055-62-8 |
---|---|
Molecular Formula | Ho2O3 |
Molecular Weight (g/mol) | 377.86 |
MDL Number | MFCD00011053 |
InChI Key | OWCYYNSBGXMRQN-UHFFFAOYSA-N |
IUPAC Name | diholmium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Ho+3].[Ho+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381428
|
Thermo Scientific Alfa Aesar
010910.03 |
1 g |
59.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15182975
|
Thermo Scientific Alfa Aesar
010910.06 |
5 g |
168.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301438
|
Thermo Scientific Alfa Aesar
010910.14 |
25 g |
664.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12055-62-8 | |
377.86 | |
OWCYYNSBGXMRQN-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Ho+3].[Ho+3] |
Ho2O3 | |
MFCD00011053 | |
diholmium(3+) trioxidandiide |
Specifications
2360°C | |
Beige | |
99.995% (REO) | |
MFCD00011053 | |
OWCYYNSBGXMRQN-UHFFFAOYSA-N | |
diholmium(3+) trioxidandiide | |
377.86 | |
Beige powder or lump | |
Holmium(III) oxide |
12055-62-8 | |
Powder | |
Ho2O3 | |
Insoluble in water. Soluble in perchloric acids. | |
[O--].[O--].[O--].[Ho+3].[Ho+3] | |
377.86 | |
Odorless | |
8.36g/cm3 |
Safety and Handling
missing translation for 'einecsNumber' : 235-015-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only