Learn More
Thermo Scientific™ Holmium Oxide Solution Verification Standard
Thermo Scientific™ NIST™-traceable Holmium Oxide Solution Standard verifies wavelength accuracy of UV-Visible spectrophotometers.
Brand:  Thermo Scientific™ 222-226400
Product Code. 11823654
  • 1540.00 € / Each
Explore more special offers
Alert:
This standard is qualified for use on instruments having a spectral bandwidth of <3nm.
Description
Description
Thermo Scientific NIST™-traceable Holmium Oxide Solution Standard verifies wavelength accuracy of UV-Visible spectrophotometers.
Holmium Oxide in Perchloric Acid Solution is a preferred standard for verifying the wavelength accuracy of a spectrophotometer according to international pharmacopoeia guidelines.
  • Permanently sealed in a quartz cuvette allowing it to be used into the deep UV range
  • Exhibits sharp, stable peaks within the range of 219 to 650nm—can be easily correlated to wavelengths of the peak maxima
  • Readings observed for each peak are verified against expected values on the certificate supplied with the standard
  • Readings not within the expected range may be indicative of an instrument performance error
This standard is qualified for use on instruments having a spectral bandwidth of <3nm.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIST/NPL Traceable
Wavelength Standard; Verification Standard
Holmium Oxide in Perchloric Acid Solution
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Holmium Oxide Solution Verification Standard > Wavelength Standard; Verification Standard