missing translation for 'onlineSavingsMsg'
Learn More

Holmium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Código de producto. 11374258
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
50 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]

Especificaciones

Color Gray to Silver
UN Number UN3178
Quantity 2 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Holmium pieces, Distilled dendritic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'